missing translation for 'onlineSavingsMsg'
Learn More

L-Serine, 98.5%, Spectrum™ Chemical

$227.93 - $3,017.04

Chemical Identifiers

CAS 56-45-1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611387
View Documents Promotion Details
N/A 25 g Poly Pail
Each for $227.93
Only null left
 
18611386
View Documents Promotion Details
N/A 100 g Poly Pail
Each for $466.14
Only null left
 
18611385
View Documents Promotion Details
N/A 1 kg Poly Pail
Each for $3,017.04
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56-45-1

Specifications

56-45-1
10 ppm
+13.6° to +15.6°
Reagent
1
0.003
98.5%
L-Serine
Videos
SDS
Documents

Documents

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.