missing translation for 'onlineSavingsMsg'
Learn More

L-Selenomethionine 98.0+%, TCI America™

Catalog No. S04421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04421G 1 g
S04425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S04421G Supplier TCI America Supplier No. S04421G

Chemical Identifiers

CAS 3211-76-5
Molecular Formula C5H11NO2Se
Molecular Weight (g/mol) 196.12
MDL Number MFCD00037210
InChI Key RJFAYQIBOAGBLC-BYPYZUCNSA-N
Synonym l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid
PubChem CID 105024
ChEBI CHEBI:62621
IUPAC Name (2S)-2-amino-4-(methylselanyl)butanoic acid
SMILES C[Se]CC[C@H](N)C(O)=O

Specifications

Melting Point 267°C
Color White-Yellow
Quantity 1 g
UN Number 3283
Formula Weight 196.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Selenomethionine
RTECSNumber EK7713840
TSCA No
Recommended Storage Freezer