missing translation for 'onlineSavingsMsg'
Learn More

L-Ribose 98.0+%, TCI America™

Catalog No. R00685G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00685G 5 g
R00681G 1 g
R006825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R00685G Supplier TCI America Supplier No. R00685G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24259-59-4 | Formula: C5H10O5 | Weight: 150.13 g/mol

Chemical Identifiers

CAS 24259-59-4
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00167010
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
PubChem CID 90428
ChEBI CHEBI:47015
IUPAC Name (3S,4R,5S)-5-(hydroxymethyl)oxolane-2,3,4-triol
SMILES OC[C@@H]1OC(O)[C@@H](O)[C@H]1O

Specifications

Melting Point 90°C
Color White
Quantity 5 g
Formula Weight 150.13
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-Ribose
EINECSNumber (5)-6838
TSCA No