missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Ribose, 99%, Thermo Scientific Chemicals

Catalog No. p-7025570
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2111703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2111703 Supplier Thermo Scientific Chemicals Supplier No. B2111703

L-Ribose is a potential starting material for the synthesis of many L-nucleoside-based pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Ribose is a potential starting material for the synthesis of many L-nucleoside-based pharmaceutical compounds.

Solubility
Soluble in water (100 mg/ml).

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 24259-59-4
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00167010
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
PubChem CID 90428
ChEBI CHEBI:47015
IUPAC Name (2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES OC[C@@H]1OC(O)[C@@H](O)[C@H]1O

Specifications

Melting Point 85°C to 88°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water (100mg/ml).
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material L-(+)-Ribose
EINECSNumber 246-110-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only