missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ribose, 99%

Catalog No. AC257910050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC257910050 5 g
AC257910250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC257910050 Supplier Thermo Scientific Chemicals Supplier No. 257910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 24259-59-4 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24259-59-4
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00167010
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
PubChem CID 90428
ChEBI CHEBI:47015
IUPAC Name (2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES OC[C@@H]1OC(O)[C@@H](O)[C@H]1O

Specifications

Melting Point 84.0°C to 86.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Quantity 5 g
Beilstein 01,859
Specific Rotation Condition + 20.00 (20.00°C c=2,water)
Specific Rotation + 20.00
Solubility Information (10% in water) Clear colorless
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material L(+)-Ribose
Show More Show Less
EINECSNumber 246-110-4

RUO – Research Use Only