missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Rhamnose Monohydrate 98.0+%, TCI America™

Catalog No. p-7135661
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00135G 5 g
R001325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R00135G Supplier TCI America Supplier No. R00135G

Chemical Identifiers

CAS 10030-85-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00149363,MFCD00136036
InChI Key SHZGCJCMOBCMKK-HGVZOGFYSA-N
Synonym l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
PubChem CID 20849066
IUPAC Name (2R,3R,4R,5R,6S)-6-methyloxane-2,3,4,5-tetrol
SMILES C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 93°C
Color White
Quantity 5 g
Formula Weight 164.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-(+)-Rhamnose Monohydrate