missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Rhamnose, monohydrate, ∽99%, MP Biomedicals™

Supplier:  MP Biomedicals Inc 0210280925

 View more versions of this product

Catalog No. ICN10280925


Only null left

Description

Description

Specifications

Specifications

L-(+)-Rhamnose, monohydrate
White
∽ 99%
MFCD00149363,MFCD00136036
l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
164.16
182.17
Solid
10030-85-0
1.47g/mL
C6H12O5
25 g
SHZGCJCMOBCMKK-HGVZOGFYSA-N
(2R,3R,4R,5R,6S)-6-methyloxane-2,3,4,5-tetrol
20849066
∼99%

Chemical Identifiers

10030-85-0
164.16
SHZGCJCMOBCMKK-HGVZOGFYSA-N
20849066
C6H12O5
MFCD00149363,MFCD00136036
l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Promotions

Promotions