missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Rhamnose, monohydrate, ∽99%, MP Biomedicals™

Catalog No. ICN10280980
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10280980 100 g
ICN10280910 10 g
ICN10280925 25 g
ICN10280950 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10280980 Supplier MP Biomedicals Inc Supplier No. 0210280980

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10030-85-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00149363,MFCD00136036
InChI Key SHZGCJCMOBCMKK-HGVZOGFYSA-N
Synonym l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
PubChem CID 20849066
SMILES C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Density 1.47g/mL
Assay Percent Range ∽ 99%
Quantity 100 g
IUPAC Name (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate
Formula Weight 182.17
Percent Purity ∼99%
Grade Not Available
Physical Form Solid
Chemical Name or Material L-(+)-Rhamnose, monohydrate
Recommended Storage Store at room temperature.