missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Rhamnose monohydrate, 99%

CAS: 10030-85-0 | C6H12O5 | 164.16 g/mol

Supplier:  Thermo Scientific Chemicals 174080250

 View more versions of this product

Catalog No. AC174080250


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

L(+)-Rhamnose monohydrate
91.0°C to 93.0°C
0.2% max.
98.5% min. (ELSD) (HPLC)
C6H12O5
25 g
+ 8.20
Solubility in water: 300g/L (20°C). Other solubilities: soluble in ethanol and methanol
C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
164.16
182.17
Crystalline Powder or Crystals
10030-85-0
White
Authentic
Glass bottle
MFCD00149363,MFCD00136036
+ 8.20 (20.00°C c=4.5,H2O)
l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
SHZGCJCMOBCMKK-HGVZOGFYSA-N
(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate
20849066
99%

Chemical Identifiers

10030-85-0
164.16
SHZGCJCMOBCMKK-HGVZOGFYSA-N
20849066
C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O
C6H12O5
MFCD00149363,MFCD00136036
l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
(2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only