missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-butyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

$53.26 - $166.42

Chemical Identifiers

CAS 5497-76-7
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00153459
InChI Key IUUYANMOEMBTBV-FJXQXJEOSA-N
Synonym h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl
PubChem CID 6453107
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES CC(C)(C)OC(=O)C1CCCN1.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1948903
View Documents
Thermo Scientific Chemicals
L1948903
1 g
Each for $53.26
Only null left
 
AAL1948906
View Documents
Thermo Scientific Chemicals
L1948906
5 g
Each for $166.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5497-76-7
207.698
IUUYANMOEMBTBV-FJXQXJEOSA-N
6453107
CC(C)(C)OC(=O)C1CCCN1.Cl
C9H18ClNO2
MFCD00153459
h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl
tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride

Specifications

110°C to 112°C
C9H18ClNO2
3596764
IUUYANMOEMBTBV-FJXQXJEOSA-N
tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
207.698
−31° (c=0.5 in Water)
≥98%
5497-76-7
MFCD00153459
h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl
CC(C)(C)OC(=O)C1CCCN1.Cl
1 g
6453107
207.7
L-Proline tert-butyl ester hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-835-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only