missing translation for 'onlineSavingsMsg'
Learn More

L-Proline tert-butyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. AAL1948903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1948903 1 g
AAL1948906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1948903 Supplier Thermo Scientific Chemicals Supplier No. L1948903
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5497-76-7
Molecular Formula C9H18ClNO2
Molecular Weight (g/mol) 207.698
MDL Number MFCD00153459
InChI Key IUUYANMOEMBTBV-FJXQXJEOSA-N
Synonym h-pro-otbu.hcl, l-proline tert-butyl ester hydrochloride, tert-butyl l-prolinate hydrochloride, l-proline tert-butyl ester hcl, pro-otbu hcl, h-pro-otbu hcl, h-pro-otbu hydrochloride, l-proline t-butyl ester hydrochloride, tert-butyl 2s-pyrrolidine-2-carboxylate hydrochloride, pro-otbu.hcl
PubChem CID 6453107
IUPAC Name tert-butyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
SMILES CC(C)(C)OC(=O)C1CCCN1.Cl

Specifications

Melting Point 110°C to 112°C
Beilstein 3596764
Quantity 1 g
Formula Weight 207.7
Optical Rotation −31° (c=0.5 in Water)
Percent Purity ≥98%
Chemical Name or Material L-Proline tert-butyl ester hydrochloride
EINECSNumber 226-835-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only