missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Proline Methyl Ester Hydrochloride 98.0+%, TCI America™
$93.03 - $341.75
Chemical Identifiers
| CAS | 2133-40-6 |
|---|---|
| Molecular Formula | C6H12NO2 |
| Molecular Weight (g/mol) | 130.17 |
| MDL Number | MFCD00012708 |
| InChI Key | BLWYXBNNBYXPPL-YFKPBYRVSA-O |
| Synonym | l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride |
| PubChem CID | 2733200 |
| IUPAC Name | (2S)-2-(methoxycarbonyl)pyrrolidin-1-ium |
| SMILES | COC(=O)[C@@H]1CCC[NH2+]1 |
Chemical Identifiers
| C6H12NO2 | |
| MFCD00012708 | |
| l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride | |
| (2S)-2-(methoxycarbonyl)pyrrolidin-1-ium |
Specifications
| 73°C | |
| 55°C | |
| C6H12NO2 | |
| l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride | |
| COC(=O)[C@@H]1CCC[NH2+]1 | |
| 130.17 | |
| 165.62 | |
| Crystalline Powder |
| 2133-40-6 | |
| White | |
| MFCD00012708 | |
| BLWYXBNNBYXPPL-YFKPBYRVSA-O | |
| (2S)-2-(methoxycarbonyl)pyrrolidin-1-ium | |
| 2733200 | |
| ≥98.0% (N) | |
| L-Proline Methyl Ester Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator