accessibility menu, dialog, popup

UserName

CH3OO(S)H2N

CAS RN 2133-40-6

IUPAC Name: (2S)-2-(methoxycarbonyl)pyrrolidin-1-ium
Synonyms: (2S)-2-(methoxycarbonyl)pyrrolidin-1-ium
Molecular Weight (g/mol): 130.17
Molecular Formula: C6H12NO2
InChi Key: BLWYXBNNBYXPPL-YFKPBYRVSA-O
SMILES: COC(=O)[C@@H]1CCC[NH2+]1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)