missing translation for 'onlineSavingsMsg'
Learn More

L-Proline methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. p-7047469
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1929003 1 g
AAL1929006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1929003 Supplier Thermo Scientific Chemicals Supplier No. L1929003

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2133-40-6
Molecular Formula C6H12NO2
Molecular Weight (g/mol) 130.17
MDL Number MFCD00012708
InChI Key BLWYXBNNBYXPPL-YFKPBYRVSA-O
Synonym l-proline methyl ester hydrochloride, h-pro-ome.hcl, methyl l-prolinate hydrochloride, methyl 2s-pyrrolidine-2-carboxylate hydrochloride, h-l-pro-ome hcl, s-methyl pyrrolidine-2-carboxylate hydrochloride, unii-7lwl7p890m, proline, methyl ester, hydrochloride, methyl l-prolinate hcl, methyl 2s pyrrolidine-2-carboxylate, chloride
PubChem CID 2733200
SMILES COC(=O)[C@@H]1CCC[NH2+]1

Specifications

Melting Point 71°C to 78°C
Beilstein 3596045
IUPAC Name methyl (2S)-pyrrolidine-2-carboxylate;hydrochloride
Quantity 1 g
Formula Weight 165.62
Optical Rotation −33° (c=1 in Water)
Percent Purity ≥98%
Sensitivity Hygroscopic
Chemical Name or Material L-Proline methyl ester hydrochloride
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 218-363-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only