missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Proline Benzyl Ester Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P172725G
Specifications
| L-Proline Benzyl Ester Hydrochloride | |
| 144°C | |
| C12H16ClNO2 | |
| 25 g | |
| NEDMOHHWRPHBAL-MERQFXBCSA-N | |
| benzyl (2S)-pyrrolidine-2-carboxylate;hydrochloride | |
| 2733413 | |
| ≥98.0% (HPLC,T) |
| 16652-71-4 | |
| White-Yellow | |
| MFCD00039002 | |
| l-proline benzyl ester hydrochloride, h-pro-obzl.hcl, l-proline benzyl ester hcl, s-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl l-prolinate hydrochloride, proline, phenylmethyl ester, hydrochloride, proline benzyl ester hydrochloride, benzyl 2s-pyrrolidine-2-carboxylate hydrochloride, l-proline, phenylmethyl ester, hydrochloride, phenylmethyl 2s pyrrolidine-2-carboxylate, chloride | |
| C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl | |
| 241.715 | |
| 241.72 | |
| Crystalline Powder |
Chemical Identifiers
| 16652-71-4 | |
| 241.715 | |
| NEDMOHHWRPHBAL-MERQFXBCSA-N | |
| 2733413 | |
| C1CC(NC1)C(=O)OCC2=CC=CC=C2.Cl |
| C12H16ClNO2 | |
| MFCD00039002 | |
| l-proline benzyl ester hydrochloride, h-pro-obzl.hcl, l-proline benzyl ester hcl, s-benzyl pyrrolidine-2-carboxylate hydrochloride, benzyl l-prolinate hydrochloride, proline, phenylmethyl ester, hydrochloride, proline benzyl ester hydrochloride, benzyl 2s-pyrrolidine-2-carboxylate hydrochloride, l-proline, phenylmethyl ester, hydrochloride, phenylmethyl 2s pyrrolidine-2-carboxylate, chloride | |
| benzyl (2S)-pyrrolidine-2-carboxylate;hydrochloride |
Safety and Handling
TSCA : No