missing translation for 'onlineSavingsMsg'
Learn More

L-Proline, 98.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor PR1201KG

 View more versions of this product

Catalog No. 18611381


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

147-85-3
0.003
1 kg
0.985
1
Poly Pail
-84.0° to -86.3°
Reagent

Chemical Identifiers

147-85-3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.