missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Proline, 98.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Loss on Drying | 0.003 |
| Packaging | Poly Bottle |
| Quantity | 25 g |
| Specific Rotation | -84.0° to -86.3° |
| Percent Purity | 0.985 |
| Grade | Reagent |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.