missing translation for 'onlineSavingsMsg'
Learn More

L-Prolinamide 97.0+%, TCI America™

Catalog No. P13821G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13821G 1 g
P13825G 5 g
P138225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P13821G Supplier TCI America Supplier No. P13821G

Chemical Identifiers

CAS 7531-52-4
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005253
InChI Key VLJNHYLEOZPXFW-BYPYZUCNSA-O
Synonym l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
PubChem CID 111306
ChEBI CHEBI:21374
IUPAC Name (2S)-2-carbamoylpyrrolidin-1-ium
SMILES NC(=O)[C@@H]1CCC[NH2+]1

Specifications

Melting Point 99°C
Color White-Yellow
Quantity 1 g
Formula Weight 114.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Prolinamide
EINECSNumber (5)-6773
TSCA No