Learn More
L-Phenylalanyl-L-phenylalanine, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals J6004303
Description
Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes. These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes
- These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation
Specifications
| L-Phenylalanyl-L-phenylalanine | |
| White | |
| MFCD00063154 | |
| Soluble at 50mg/ml in 80% acetic acid | |
| C1=CC=C(C=C1)CC(C(=O)NC(CC2=CC=CC=C2)C(=O)O)N | |
| 1 g | |
| 6993090 | |
| 312.37 |
| 2577-40-4 | |
| C18H20N2O3 | |
| h-phe-phe-oh, l-phe-l-phe, s-2-s-2-amino-3-phenylpropanamido-3-phenylpropanoic acid, chembl55868, phe-phe, 2s-2-2s-2-amino-3-phenylpropanamido-3-phenylpropanoic acid, l-phenylalanyl-l-phenylalanine, phenylalanyl-phenylalanine, 2s-2-2s-2-amino-3-phenylpropanoyl amino-3-phenylpropanoic acid | |
| GKZIWHRNKRBEOH-HOTGVXAUSA-N | |
| (2S)-2-[[(2S)-2-amino-3-phenylpropanoyl]amino]-3-phenylpropanoic acid | |
| 312.369 | |
| CHEBI:72723 | |
| Powder |
Chemical Identifiers
| 2577-40-4 | |
| 312.369 | |
| GKZIWHRNKRBEOH-HOTGVXAUSA-N | |
| 6993090 | |
| (2S)-2-[[(2S)-2-amino-3-phenylpropanoyl]amino]-3-phenylpropanoic acid |
| C18H20N2O3 | |
| MFCD00063154 | |
| h-phe-phe-oh, l-phe-l-phe, s-2-s-2-amino-3-phenylpropanamido-3-phenylpropanoic acid, chembl55868, phe-phe, 2s-2-2s-2-amino-3-phenylpropanamido-3-phenylpropanoic acid, l-phenylalanyl-l-phenylalanine, phenylalanyl-phenylalanine, 2s-2-2s-2-amino-3-phenylpropanoyl amino-3-phenylpropanoic acid | |
| CHEBI:72723 | |
| C1=CC=C(C=C1)CC(C(=O)NC(CC2=CC=CC=C2)C(=O)O)N |
Safety and Handling
EINECSNumber : 219-930-5
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only