missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanyl-L-phenylalanine, Thermo Scientific Chemicals

Catalog No. p-7041730
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6004303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6004303 Supplier Thermo Scientific Chemicals Supplier No. J6004303

Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes. These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Evaporating 1,1,1,3,3,3-hexafluoro-2-propanol (HFIP) solutions containing diphenylalanine forms nanotubes
  • These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation

Chemical Identifiers

CAS 2577-40-4
Molecular Formula C18H20N2O3
Molecular Weight (g/mol) 312.369
MDL Number MFCD00063154
InChI Key GKZIWHRNKRBEOH-HOTGVXAUSA-N
Synonym h-phe-phe-oh, l-phe-l-phe, s-2-s-2-amino-3-phenylpropanamido-3-phenylpropanoic acid, chembl55868, phe-phe, 2s-2-2s-2-amino-3-phenylpropanamido-3-phenylpropanoic acid, l-phenylalanyl-l-phenylalanine, phenylalanyl-phenylalanine, 2s-2-2s-2-amino-3-phenylpropanoyl amino-3-phenylpropanoic acid
PubChem CID 6993090
ChEBI CHEBI:72723
IUPAC Name (2S)-2-[[(2S)-2-amino-3-phenylpropanoyl]amino]-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)CC(C(=O)NC(CC2=CC=CC=C2)C(=O)O)N

Specifications

Color White
Solubility Information Soluble at 50mg/ml in 80% acetic acid
Quantity 1 g
Formula Weight 312.37
Physical Form Powder
Chemical Name or Material L-Phenylalanyl-L-phenylalanine
EINECSNumber 219-930-5
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only