missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalaninol 98.0+%, TCI America™

Catalog No. p-7134909
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10285G 5 g
P102825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10285G Supplier TCI America Supplier No. P10285G
Only null left

Chemical Identifiers

CAS 3182-95-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00004732
InChI Key STVVMTBJNDTZBF-VIFPVBQESA-N
Synonym l-phenylalaninol, s-2-amino-3-phenyl-1-propanol, s---2-amino-3-phenyl-1-propanol, s-2-amino-3-phenylpropan-1-ol, h-phenylalaninol, h-phe-ol, 2s-2-amino-3-phenylpropan-1-ol, phenylalaninol, --l-phenylalaninol, l-phenyl alaninol
PubChem CID 447213
IUPAC Name (2S)-2-amino-3-phenylpropan-1-ol
SMILES N[C@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 94°C
Color White-Yellow
Quantity 5 g
UN Number 3259
Formula Weight 151.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Phenylalaninol
RTECSNumber UA6900000
TSCA No