accessibility menu, dialog, popup

UserName

NH2(S)OH

CAS RN 3182-95-4

IUPAC Name: (2S)-2-amino-3-phenylpropan-1-ol
Synonyms: L-phenylalaninol|2-amino-3-phenyl-1-propanol|S-phenylalaninol|(S)-2-benzylethanolamine|(R)-2-amino-3-phenylpropanol|(S)-β-aminobenzenepropanol
Molecular Weight (g/mol): 151.21
Molecular Formula: C9H13NO
InChi Key: STVVMTBJNDTZBF-VIFPVBQESA-N
SMILES: N[C@H](CO)CC1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)