missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Phenylalaninol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 94°C |
| Color | White-Yellow |
| Quantity | 5 g |
| UN Number | 3259 |
| Formula Weight | 151.21 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Phenylalaninol |
Safety and Handling
| RTECSNumber | UA6900000 |
| TSCA | No |