missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. P127825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P127825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P127825G Supplier TCI America Supplier No. P127825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7524-50-7
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00012489
InChI Key SWVMLNPDTIFDDY-UHFFFAOYNA-N
Synonym l-phenylalanine methyl ester hydrochloride, h-phe-ome.hcl, methyl l-phenylalaninate hydrochloride, s-methyl 2-amino-3-phenylpropanoate hydrochloride, unii-47hk4y94ja, h-phe-ome hcl, h-phe-ome hydrochloride, l-phenylalanine, methyl ester, hydrochloride, l-phenylalanine methyl ester hcl
PubChem CID 75736
IUPAC Name hydrogen methyl 2-amino-3-phenylpropanoate chloride
SMILES [H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1

Specifications

Quantity 25 g
Formula Weight 215.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Phenylalanine Methyl Ester Hydrochloride
EINECSNumber (9)-1610
TSCA No