accessibility menu, dialog, popup

UserName

CAS RN 7524-50-7

IUPAC Name: hydrogen methyl 2-amino-3-phenylpropanoate chloride
Synonyms: hydrogen methyl 2-amino-3-phenylpropanoate chloride
Molecular Weight (g/mol): 215.68
Molecular Formula: C10H14ClNO2
InChi Key: SWVMLNPDTIFDDY-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)