missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine methyl ester hydrochloride, 98%

Catalog No. p-181809
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130320100 10 g Glass bottle
AC130320500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC130320100 Supplier Thermo Scientific Chemicals Supplier No. 130320100

CAS: 7524-50-7 | C10H14ClNO2 | 215.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7524-50-7
Molecular Formula C10H14ClNO2
Molecular Weight (g/mol) 215.68
MDL Number MFCD00012489
InChI Key SWVMLNPDTIFDDY-UHFFFAOYNA-N
Synonym l-phenylalanine methyl ester hydrochloride, h-phe-ome.hcl, methyl l-phenylalaninate hydrochloride, s-methyl 2-amino-3-phenylpropanoate hydrochloride, unii-47hk4y94ja, h-phe-ome hcl, h-phe-ome hydrochloride, l-phenylalanine, methyl ester, hydrochloride, l-phenylalanine methyl ester hcl
PubChem CID 75736
SMILES [H+].[Cl-].COC(=O)C(N)CC1=CC=CC=C1

Specifications

Melting Point 156.0°C to 160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula C6H5CH2CH(NH2)CO2CH3·HCl
Quantity 10 g
Beilstein 14,499
Specific Rotation Condition + 37.00 (20.00°C c=2,C2H5OH)
Specific Rotation + 37.00
IUPAC Name methyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
Formula Weight 215.68
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material L-Phenylalanine methyl ester hydrochloride, 98%
Show More Show Less
EINECSNumber 231-383-4

RUO – Research Use Only