missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ L-Phenylalanine ethyl ester hydrochloride, 99%

$122.92 - $516.47

Chemical Identifiers

CAS 3182-93-2
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.70
MDL Number MFCD00012507
InChI Key FPFQPLFYTKMCHN-PPHPATTJSA-N
Synonym l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
PubChem CID 165085
IUPAC Name ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC205430100
View Documents Promotion Details
Thermo Scientific™
205430100
10g Plastic bottle
Each for $122.92
Only null left
 
AC205430500
View Documents Promotion Details
Thermo Scientific™
205430500
50g Plastic bottle
Each for $516.47
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3182-93-2
229.70
FPFQPLFYTKMCHN-PPHPATTJSA-N
165085
[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
C11H16ClNO2
MFCD00012507
l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride

Specifications

3182-93-2
99%
C6H5CH2CH(NH2)CO2C2H5·HCl
14, 497
+ 33.7
FPFQPLFYTKMCHN-PPHPATTJSA-N
ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
165085
99%
Authentic
C11H16ClNO2
MFCD00012507
+33.70° (21°C c=2,C2H5OH)
l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
[H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1
229.70
229.71
L-Phenylalanine ethyl ester hydrochloride, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 221-673-9

SDS
Documents

Documents

RUO – Research Use Only