missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Phenylalanine ethyl ester hydrochloride, 99%

Catalog No. AC205430500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC205430500 50 g Plastic bottle
AC205430100 10 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC205430500 Supplier Thermo Scientific Chemicals Supplier No. 205430500

CAS: 3182-93-2 | C11H16ClNO2 | 229.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3182-93-2
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.70
MDL Number MFCD00012507
InChI Key FPFQPLFYTKMCHN-PPHPATTJSA-N
Synonym l-phenylalanine ethyl ester hydrochloride, h-phe-oet.hcl, ethyl l-phenylalaninate hydrochloride, s-ethyl 2-amino-3-phenylpropanoate hydrochloride, ethyl 2s-2-amino-3-phenylpropanoate hydrochloride, l-phenylalanine, ethyl ester, hydrochloride, l-phenylalanine ethylester, ethyl l-phenylalaninate hcl, phenylalanine ethyl ester hydrochloride, phe-oet.hcl
PubChem CID 165085
IUPAC Name ethyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES [H+].[Cl-].CCOC(=O)[C@@H](N)CC1=CC=CC=C1

Specifications

Melting Point 154°C to 157°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula C6H5CH2CH(NH2)CO2C2H5·HCl
Quantity 50 g
Beilstein 14, 497
Specific Rotation Condition +33.70° (21°C c=2,C2H5OH)
Specific Rotation + 33.7
Formula Weight 229.71
Percent Purity 99%
Physical Form Fine Needle-like Crystalline Solid
Chemical Name or Material L-Phenylalanine ethyl ester hydrochloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 221-673-9

RUO – Research Use Only