missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Phenylalanine, Cell Culture Reagent

Catalog No. AAJ6392522
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6392522 100 g
AAJ6392530 250 g
AAJ6392536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6392522 Supplier Thermo Scientific Chemicals Supplier No. J6392522

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol

The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The primary use of L-phenylalanine in the body is in the production of tyrosine, another amino acid. It is an essential amino acid, which is biologically converted into L-tyrosine, another one of the DNA-encoded amino acids, which in turn is converted to L-DOPA and further converted into dopamine, norepinephrine, and epinephrine. L-Phenylalanine is produced for medical, feed, and nutritional applications such as in the preparation of Aspartame.

Solubility
Soluble in water. Aqueous solubility increases with warming. Also soluble in methanol, ethanol or 1M NH{4}OH

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064227
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Color White
Beilstein 1910408
Merck Index 14,7271
Solubility Information Soluble in water; Aqueous solubility increases with warming; Also soluble in methanol,ethanol or 1M NH4OH
IUPAC Name 2-amino-3-phenylpropanoic acid
Quantity 100 g
Formula Weight 165.19
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Phenylalanine
EINECSNumber 200-568-1
RTECSNumber AY7535000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only