missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Phenylalanine Benzyl Ester Hydrochloride 98.0+%, TCI America™
$59.69 - $240.64
Chemical Identifiers
| CAS | 2462-32-0 |
|---|---|
| Molecular Formula | C16H18ClNO2 |
| Molecular Weight (g/mol) | 291.78 |
| MDL Number | MFCD00043249 |
| InChI Key | CEXFHIYDTRNBJD-RSAXXLAASA-N |
| Synonym | l-phenylalanine benzyl ester hydrochloride, h-phe-obzl.hcl, h-phe-obzl hcl, l-phenylalanine benzyl ester hcl, s-benzyl 2-amino-3-phenylpropanoate hydrochloride, phenylalanine, phenylmethyl ester, hydrochloride, benzyl 2s-2-amino-3-phenylpropanoate hydrochloride, phenylmethyl 2s-2-amino-3-phenylpropanoate, chloride, pubchem19007, benzyl phenylalaninate hydrochloride |
| PubChem CID | 12636208 |
| IUPAC Name | benzyl (2S)-2-amino-3-phenylpropanoate hydrochloride |
| SMILES | Cl.N[C@@H](CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 2462-32-0 | |
| 291.78 | |
| CEXFHIYDTRNBJD-RSAXXLAASA-N | |
| 12636208 | |
| Cl.N[C@@H](CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 |
| C16H18ClNO2 | |
| MFCD00043249 | |
| l-phenylalanine benzyl ester hydrochloride, h-phe-obzl.hcl, h-phe-obzl hcl, l-phenylalanine benzyl ester hcl, s-benzyl 2-amino-3-phenylpropanoate hydrochloride, phenylalanine, phenylmethyl ester, hydrochloride, benzyl 2s-2-amino-3-phenylpropanoate hydrochloride, phenylmethyl 2s-2-amino-3-phenylpropanoate, chloride, pubchem19007, benzyl phenylalaninate hydrochloride | |
| benzyl (2S)-2-amino-3-phenylpropanoate hydrochloride |
Specifications
| 2462-32-0 | |
| White-Yellow | |
| MFCD00043249 | |
| l-phenylalanine benzyl ester hydrochloride, h-phe-obzl.hcl, h-phe-obzl hcl, l-phenylalanine benzyl ester hcl, s-benzyl 2-amino-3-phenylpropanoate hydrochloride, phenylalanine, phenylmethyl ester, hydrochloride, benzyl 2s-2-amino-3-phenylpropanoate hydrochloride, phenylmethyl 2s-2-amino-3-phenylpropanoate, chloride, pubchem19007, benzyl phenylalaninate hydrochloride | |
| Cl.N[C@@H](CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1 | |
| 291.78 | |
| 291.78 | |
| Crystalline Powder |
| 203°C | |
| C16H18ClNO2 | |
| 5 g | |
| CEXFHIYDTRNBJD-RSAXXLAASA-N | |
| benzyl (2S)-2-amino-3-phenylpropanoate hydrochloride | |
| 12636208 | |
| ≥98.0% (HPLC,N) | |
| L-Phenylalanine Benzyl Ester Hydrochloride |
Safety and Handling
TSCA : No