accessibility menu, dialog, popup

UserName

(S)OONH2HCl

CAS RN 2462-32-0

IUPAC Name: benzyl (2S)-2-amino-3-phenylpropanoate hydrochloride
Synonyms: benzyl (2S)-2-amino-3-phenylpropanoate hydrochloride
Molecular Weight (g/mol): 291.78
Molecular Formula: C16H18ClNO2
InChi Key: CEXFHIYDTRNBJD-RSAXXLAASA-N
SMILES: Cl.N[C@@H](CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)