missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine benzyl ester hydrochloride, 98%

Catalog No. AAH2840714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2840714 25 g
AAH2840706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2840714 Supplier Thermo Scientific Chemicals Supplier No. H2840714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2462-32-0 | C16H18ClNO2 | 291.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2462-32-0
Molecular Formula C16H18ClNO2
Molecular Weight (g/mol) 291.78
MDL Number MFCD00043249
InChI Key CEXFHIYDTRNBJD-RSAXXLAASA-N
Synonym l-phenylalanine benzyl ester hydrochloride, h-phe-obzl.hcl, h-phe-obzl hcl, l-phenylalanine benzyl ester hcl, s-benzyl 2-amino-3-phenylpropanoate hydrochloride, phenylalanine, phenylmethyl ester, hydrochloride, benzyl 2s-2-amino-3-phenylpropanoate hydrochloride, phenylmethyl 2s-2-amino-3-phenylpropanoate, chloride, pubchem19007, benzyl phenylalaninate hydrochloride
PubChem CID 12636208
IUPAC Name benzyl (2S)-2-amino-3-phenylpropanoate;hydrochloride
SMILES Cl.N[C@@H](CC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 197°C to 200°C
Quantity 25 g
Formula Weight 291.77
Percent Purity 98%
Chemical Name or Material L-Phenylalanine benzyl ester hydrochloride
EINECSNumber 219-558-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only