missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Phenylalanine, 98.5+%

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol

Supplier:  Thermo Scientific Chemicals 130311000

 View more versions of this product

Catalog No. AC130311000


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

270.0°C to 275.0°C
63-91-2
White
Authentic
0.3% max. (105°C, 3 hrs)
C9H11NO2
C6H5CH2CH(NH2)CO2H
− 34.10
l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
COLNVLDHVKWLRT-UHFFFAOYNA-N
(2S)-2-amino-3-phenylpropanoic acid
165.19
6140
165.19
Crystalline Powder or Crystals
L-Phenylalanine
5.4 to 6.0 (1% aq. soln.)
20ppm max.
10ppm max.
98.5+%
Plastic Bottle
MFCD00064227
0.1% max.
Solubility in water: 27g/L (20°C). Other solubilities: very slightly soluble in methanol, ethanol
NC(CC1=CC=CC=C1)C(O)=O
− 34.10 (20.00°C c=2,water,dry basis)
100 g
CHEBI:17295
≥98.5%

Chemical Identifiers

63-91-2
165.19
COLNVLDHVKWLRT-UHFFFAOYNA-N
6140
NC(CC1=CC=CC=C1)C(O)=O
C9H11NO2
MFCD00064227
l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
CHEBI:17295
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only