missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine, 98.5-101.0%, specified according to the requirements of USP

Numéro de catalogue. p-7077449
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AC453930250 25 g
AC453931000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC453930250 Fournisseur Thermo Scientific Chemicals Code fournisseur 453930250
Il en reste null

CAS: 63-91-2 | C9H11NO2 | 165.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
SMILES NC(CC1=CC=CC=C1)C(O)=O

Spécifications

Melting Point 270.0°C to 275.0°C
pH 5.4 to 6.0 (1% solution)
Chloride 0.05% max.
Heavy Metals 15ppm max.
Infrared Spectrum Authentic
Iron (Fe) 0.003% max.
Loss on Drying 0.3% max. (105°C, 3 hrs)
Linear Formula C6H5CH2CH(NH2)CO2H
Specific Rotation −34.10
Sulfate 0.03% max.
Sulfated Ash 0.4% max.
Solubility Information Solubility in water: 27g/L (20°C). Other solubilities: very slightly soluble in methanol, ethanol
IUPAC Name 2-amino-3-phenylpropanoic acid
Specific Rotation Condition −34.10 (20.00°C c=2,water,dry basis)
Quantity 25 g
Formula Weight 165.19
Chemical Name or Material L-Phenylalanine, 98.5-101.0%
Afficher plus de résultats Afficher moins de résultats

RUO – Research Use Only