missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalaninamide Hydrochloride 98.0+%, TCI America™

Supplier:  TCI America P17995G

Catalog No. P17995G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

L-Phenylalaninamide Hydrochloride
234°C
C9H13ClN2O
5 g
KLHLGTPNBQXSJT-QRPNPIFTSA-N
(2S)-2-amino-3-phenylpropanamide;hydrochloride
11953719
≥98.0% (HPLC,N)
65864-22-4
White-Yellow
MFCD00039083
l-phenylalaninamide hydrochloride, h-phe-nh2.hcl, h-phe-nh2 hcl, l-phenylalanine amide hydrochloride, phenylalanine amide hydrochloride, h-phe-nh2hcl, 2s-2-amino-3-phenylpropanamide hydrochloride, c9h12n2o.hcl, phenh2hcl, h-phe-nh hcl
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
200.666
200.67
Crystalline Powder

Chemical Identifiers

65864-22-4
200.666
KLHLGTPNBQXSJT-QRPNPIFTSA-N
11953719
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
C9H13ClN2O
MFCD00039083
l-phenylalaninamide hydrochloride, h-phe-nh2.hcl, h-phe-nh2 hcl, l-phenylalanine amide hydrochloride, phenylalanine amide hydrochloride, h-phe-nh2hcl, 2s-2-amino-3-phenylpropanamide hydrochloride, c9h12n2o.hcl, phenh2hcl, h-phe-nh hcl
(2S)-2-amino-3-phenylpropanamide;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions