accessibility menu, dialog, popup

UserName

CAS RN 65864-22-4

ONH2H2NHCl

CAS RN 65864-22-4

IUPAC Name: 2-amino-3-phenylpropanamide hydrochloride
Synonyms: phenylalanine amide hydrochloride
Molecular Weight (g/mol): 200.67
Molecular Formula: C9H13ClN2O
InChi Key: KLHLGTPNBQXSJT-UHFFFAOYNA-N
SMILES: Cl.NC(CC1=CC=CC=C1)C(N)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)