missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalaninamide hydrochloride, 97%, Thermo Scientific Chemicals

$451.92 - $1488.15

Chemical Identifiers

CAS 65864-22-4
Molecular Formula C9H13ClN2O
Molecular Weight (g/mol) 200.666
MDL Number MFCD00039083
InChI Key KLHLGTPNBQXSJT-QRPNPIFTSA-N
PubChem CID 11953719
IUPAC Name (2S)-2-amino-3-phenylpropanamide;hydrochloride
SMILES C1=CC=C(C=C1)CC(C(=O)N)N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6550614
View Documents
Thermo Scientific Chemicals
H6550614
25 g
Each for $451.92
Only null left
 
AAH6550622
View Documents
Thermo Scientific Chemicals
H6550622
100 g
Each for $1,488.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

65864-22-4
200.666
KLHLGTPNBQXSJT-QRPNPIFTSA-N
(2S)-2-amino-3-phenylpropanamide;hydrochloride
C9H13ClN2O
MFCD00039083
11953719
C1=CC=C(C=C1)CC(C(=O)N)N.Cl

Specifications

231°C to 237°C
C9H13ClN2O
3915662
KLHLGTPNBQXSJT-QRPNPIFTSA-N
(2S)-2-amino-3-phenylpropanamide;hydrochloride
200.666
200.67
Powder
65864-22-4
MFCD00039083
l-phenylalaninamide hydrochloride, h-phe-nh2.hcl, h-phe-nh2 hcl, l-phenylalanine amide hydrochloride, phenylalanine amide hydrochloride, h-phe-nh2hcl, 2s-2-amino-3-phenylpropanamide hydrochloride, c9h12n2o.hcl, phenh2hcl, h-phe-nh hcl
C1=CC=C(C=C1)CC(C(=O)N)N.Cl
25 g
11953719
97%
L-Phenylalaninamide hydrochloride
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only