missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Ornithine hydrochloride

CAS: 3184-13-2 | C5H13ClN2O2 | 168.621 g/mol

Supplier:  Thermo Scientific Chemicals J6024118

 View more versions of this product

Catalog No. AAJ6024118


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

∽238°C (decomposition)
3184-13-2
C5H13ClN2O2
3625847
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
C(CC(C(=O)O)N)CN.Cl
50 g
76654
168.62
Crystalline Powder
L-Ornithine hydrochloride
White
MFCD00064562
14,6874
GGTYBZJRPHEQDG-WCCKRBBISA-N
(2S)-2,5-diaminopentanoic acid;hydrochloride
168.621
+23° (c=5 in 5N HCl)
Cell Culture Reagent

Chemical Identifiers

3184-13-2
168.621
GGTYBZJRPHEQDG-WCCKRBBISA-N
76654
C(CC(C(=O)O)N)CN.Cl
C5H13ClN2O2
MFCD00064562
l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
(2S)-2,5-diaminopentanoic acid;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 221-678-6

RTECSNumber : RM2985000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only