missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Ornithine hydrochloride
CAS: 3184-13-2 | C5H13ClN2O2 | 168.621 g/mol
Chemical Identifiers
| CAS | 3184-13-2 |
|---|---|
| Molecular Formula | C5H13ClN2O2 |
| Molecular Weight (g/mol) | 168.621 |
| MDL Number | MFCD00064562 |
| InChI Key | GGTYBZJRPHEQDG-WCCKRBBISA-N |
| Synonym | l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate |
| PubChem CID | 76654 |
| IUPAC Name | (2S)-2,5-diaminopentanoic acid;hydrochloride |
| SMILES | C(CC(C(=O)O)N)CN.Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ6024118
|
Thermo Scientific Chemicals
J6024118 |
50 g |
N/A
|
|
|||||
|
AAJ6024122
|
Thermo Scientific Chemicals
J6024122 |
100 g |
N/A
|
|
|||||
|
AAJ6024130
|
Thermo Scientific Chemicals
J6024130 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C5H13ClN2O2 | |
| MFCD00064562 | |
| l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate | |
| (2S)-2,5-diaminopentanoic acid;hydrochloride |
Specifications
| ∽238°C (decomposition) | |
| White | |
| MFCD00064562 | |
| 14,6874 | |
| GGTYBZJRPHEQDG-WCCKRBBISA-N | |
| (2S)-2,5-diaminopentanoic acid;hydrochloride | |
| 76654 | |
| +23° (c=5 in 5N HCl) | |
| Crystalline Powder |
| 3184-13-2 | |
| C5H13ClN2O2 | |
| 3625847 | |
| l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate | |
| C(CC(C(=O)O)N)CN.Cl | |
| 168.621 | |
| 168.62 | |
| Cell Culture Reagent | |
| L-Ornithine hydrochloride |
Safety and Handling
EINECSNumber : 221-678-6
RTECSNumber : RM2985000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only