missing translation for 'onlineSavingsMsg'
Learn More

L-O-Phosphoserine 98.0+%, TCI America™

Catalog No. P07735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P07735G 5 g
P077325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P07735G Supplier TCI America Supplier No. P07735G
Only null left

Chemical Identifiers

CAS 407-41-0
Molecular Formula C3H8NO6P
Molecular Weight (g/mol) 185.072
MDL Number MFCD00065935
InChI Key BZQFBWGGLXLEPQ-REOHCLBHSA-N
Synonym o-phospho-l-serine, dexfosfoserine, phosphoserine, l-o-phosphoserine, fosforina, l-sop, o-phosphoserine, l-phosphoserine, 3-phosphoserine, phosphonoserine
PubChem CID 68841
ChEBI CHEBI:15811
IUPAC Name (2S)-2-amino-3-phosphonooxypropanoic acid
SMILES C(C(C(=O)O)N)OP(=O)(O)O

Specifications

Melting Point 167°C
Quantity 5 g
Formula Weight 185.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-O-Phosphoserine