CAS RN 407-41-0
CAS RN 407-41-0
IUPAC Name:
(2S)-2-amino-3-(phosphonooxy)propanoic acid
Synonyms:
phosphoserine phosphonoserine dexfosfoserine
Molecular Weight (g/mol):
185.07
Molecular Formula:
C3H8NO6P
InChi Key:
BZQFBWGGLXLEPQ-REOHCLBHSA-N
SMILES:
N[C@@H](COP(O)(O)=O)C(O)=O
1
–
1
of
1
results
1
L-O-Phosphoserine 98.0+%, TCI America™
CAS: 407-41-0 Molecular Formula: C3H8NO6P Molecular Weight (g/mol): 185.072 MDL Number: MFCD00065935 InChI Key: BZQFBWGGLXLEPQ-REOHCLBHSA-N Synonym: o-phospho-l-serine,dexfosfoserine,phosphoserine,l-o-phosphoserine,fosforina,l-sop,o-phosphoserine,l-phosphoserine,3-phosphoserine,phosphonoserine PubChem CID: 68841 ChEBI: CHEBI:15811 IUPAC Name: (2S)-2-amino-3-phosphonooxypropanoic acid SMILES: C(C(C(=O)O)N)OP(=O)(O)O