missing translation for 'onlineSavingsMsg'
Learn More

L-Norvaline 99.0+%, TCI America™

Catalog No. N06861G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06861G 1 g
N06865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N06861G Supplier TCI America Supplier No. N06861G

Chemical Identifiers

CAS 6600-40-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064421
InChI Key SNDPXSYFESPGGJ-BYPYZUCNSA-N
Synonym l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
PubChem CID 65098
ChEBI CHEBI:18314
IUPAC Name (2S)-2-aminopentanoic acid
SMILES CCC[C@H](N)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 117.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Norvaline
EINECSNumber (2)-3889
TSCA No