accessibility menu, dialog, popup

UserName

(S)OHONH2H3C

CAS RN 6600-40-4

IUPAC Name: (2S)-2-aminopentanoic acid
Synonyms: norvaline|2-amino-pentanoic acid
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: SNDPXSYFESPGGJ-BYPYZUCNSA-N
SMILES: CCC[C@H](N)C(O)=O
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)