missing translation for 'onlineSavingsMsg'
Learn More

L-Norvaline, MP Biomedicals™

$476.47 - $476.47

Chemical Identifiers

CAS 6600-40-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064421
InChI Key SNDPXSYFESPGGJ-BYPYZUCNSA-N
Synonym l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
PubChem CID 65098
ChEBI CHEBI:18314
IUPAC Name (2S)-2-aminopentanoic acid
SMILES CCC[C@H](N)C(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10249510
View Documents
MP Biomedicals Inc
0210249510
10 g
Each for $476.47
Only null left
 
Description

Description

Specifications

Chemical Identifiers

6600-40-4
117.15
SNDPXSYFESPGGJ-BYPYZUCNSA-N
65098
(2S)-2-aminopentanoic acid
C5H11NO2
MFCD00064421
l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
CHEBI:18314
CCC[C@H](N)C(O)=O

Specifications

6600-40-4
MFCD00064421
SNDPXSYFESPGGJ-BYPYZUCNSA-N
(2S)-2-aminopentanoic acid
117.15
CHEBI:18314
Solid
C5H11NO2
l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
CCC[C@H](N)C(O)=O
10 g
65098
117.1
L-Norvaline
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 229-543-3

Recommended Storage : Store at room Temperature.

SDS
Documents

Documents