missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Norvaline, 99%

CAS: 6600-40-4 | C5H11NO2 | 117.15 g/mol

$92.47 - $193.92

Chemical Identifiers

CAS 6600-40-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00064421
InChI Key SNDPXSYFESPGGJ-BYPYZUCNSA-N
Synonym l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
PubChem CID 65098
ChEBI CHEBI:18314
IUPAC Name (2S)-2-aminopentanoic acid
SMILES CCC[C@H](N)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC226020010
SDS View Documents
Thermo Scientific Chemicals
226020010
1 g Glass Bottle
Each for $92.47
Only null left
 
AC226020050
SDS View Documents
Thermo Scientific Chemicals
226020050
5 g Glass Bottle
Each for $193.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6600-40-4
117.15
SNDPXSYFESPGGJ-BYPYZUCNSA-N
65098
(2S)-2-aminopentanoic acid
C5H11NO2
MFCD00064421
l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
CHEBI:18314
CCC[C@H](N)C(O)=O

Specifications

6600-40-4
Authentic
(TLC) not detectable
Glass Bottle
+23.5° to +25.5° (20°C, 589nm) (c=10, 6N HCl)
15, 6805
SNDPXSYFESPGGJ-BYPYZUCNSA-N
(2S)-2-aminopentanoic acid
1 g
65098
117.15
Fine Crystalline Powder
White
99%
C5H11NO2
MFCD00064421
04, 416
l-norvaline, norvaline, s-2-aminopentanoic acid, h-nva-oh, 2s-2-aminopentanoic acid, l-2-aminovaleric acid, l-2-aminopentanoic acid, norvaline, l, s-2-aminovaleric acid, l-+-norvaline
CCC[C@H](N)C(O)=O
24.50 (20.00°C c=10,6 N HCl)
117.15
CHEBI:18314
98.5 to 101.5%
L(+)-Norvaline
Videos
SDS
Documents

Documents

RUO – Research Use Only