missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Methioninol 98.0+%, TCI America™

Catalog No. M0929100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0929100MG 100 mg
M09291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0929100MG Supplier TCI America Supplier No. M0929100MG

Chemical Identifiers

CAS 2899-37-8
Molecular Formula C5H13NOS
Molecular Weight (g/mol) 135.23
MDL Number MFCD00004735
InChI Key MIQJGZAEWQQAPN-YFKPBYRVSA-N
Synonym l-methioninol, l---methioninol, h-methioninol, 2s-2-amino-4-methylsulfanyl butan-1-ol, s---methioninol, s-2-amino-4-methylthio butan-1-ol, d-s---methionanol, methioninol, 2s-2-amino-4-methylsulfanylbutan-1-ol, s-2-amino-4-methylsulfanyl-butan-1-ol
PubChem CID 2724404
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butan-1-ol
SMILES CSCC[C@H](N)CO

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 135.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Methioninol
TSCA No
Recommended Storage Refrigerator