accessibility menu, dialog, popup

UserName

CAS RN 2899-37-8

IUPAC Name: (2S)-2-amino-4-(methylsulfanyl)butan-1-ol
Synonyms: (2S)-2-amino-4-(methylsulfanyl)butan-1-ol| L-METHIONINOL
Molecular Weight (g/mol): 135.23
Molecular Formula: C5H13NOS
InChi Key: MIQJGZAEWQQAPN-YFKPBYRVSA-N
SMILES: CSCC[C@H](N)CO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)