missing translation for 'onlineSavingsMsg'
Learn More

L-Methioninol, 99+%

CAS: 2899-37-8 | C5H13NOS | 135.23 g/mol

$160.53 - $160.53

Chemical Identifiers

CAS 2899-37-8
Molecular Formula C5H13NOS
Molecular Weight (g/mol) 135.23
MDL Number MFCD00004735
InChI Key MIQJGZAEWQQAPN-YFKPBYRVSA-N
Synonym l-methioninol, l---methioninol, h-methioninol, 2s-2-amino-4-methylsulfanyl butan-1-ol, s---methioninol, s-2-amino-4-methylthio butan-1-ol, d-s---methionanol, methioninol, 2s-2-amino-4-methylsulfanylbutan-1-ol, s-2-amino-4-methylsulfanyl-butan-1-ol
PubChem CID 2724404
SMILES CSCC[C@H](N)CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229230010
SDS View Documents
Thermo Scientific Chemicals
229230010
1 g Glass bottle
Each for $160.53
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2899-37-8
135.23
MIQJGZAEWQQAPN-YFKPBYRVSA-N
2724404
C5H13NOS
MFCD00004735
l-methioninol, l---methioninol, h-methioninol, 2s-2-amino-4-methylsulfanyl butan-1-ol, s---methioninol, s-2-amino-4-methylthio butan-1-ol, d-s---methionanol, methioninol, 2s-2-amino-4-methylsulfanylbutan-1-ol, s-2-amino-4-methylsulfanyl-butan-1-ol
CSCC[C@H](N)CO

Specifications

Authentic
2899-37-8
-11.5° to -13.5° (20°C, 589nm) (c=1.4, ethanol)
1 g
99+%
MFCD00004735
MIQJGZAEWQQAPN-YFKPBYRVSA-N
(2S)-2-amino-4-(methylsulfanyl)butan-1-ol
2724404
≥99.0%
White
Glass bottle
−12.50° (20°C c=1.4,C2H5OH)
Crystalline Low Melting Solid
C5H13NOS
l-methioninol, l---methioninol, h-methioninol, 2s-2-amino-4-methylsulfanyl butan-1-ol, s---methioninol, s-2-amino-4-methylthio butan-1-ol, d-s---methionanol, methioninol, 2s-2-amino-4-methylsulfanylbutan-1-ol, s-2-amino-4-methylsulfanyl-butan-1-ol
CSCC[C@H](N)CO
135.23
135.22
L-Methioninol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

RUO – Research Use Only