missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ L-Methionine methyl ester hydrochloride, 98%

Supplier:  Thermo Scientific™ 229350500

 View more versions of this product

Catalog No. AC229350500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

L-Methionine methyl ester hydrochloride
2491-18-1
97.5 to 102.5% (Argentometry)
C6H14ClNO2S
MFCD00012491,MFCD00067540
+26° (24°C c=5,H2O)
h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
MEVUPUNLVKELNV-UHFFFAOYNA-N
methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
11435579
98%
99+%
Authentic
Glass bottle
CH3SCH2CH2CH(NH2)CO2CH3·HCl
50g
+ 26
(5% in methanol) Clear colorless to light yellow
[H+].[Cl-].COC(=O)C(N)CCSC
199.69
199.7

Chemical Identifiers

2491-18-1
199.69
MEVUPUNLVKELNV-UHFFFAOYNA-N
11435579
[H+].[Cl-].COC(=O)C(N)CCSC
C6H14ClNO2S
MFCD00012491,MFCD00067540
h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
methyl (2S)-2-amino-4-methylsulfanylbutanoate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-651-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only