missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine Methyl Ester Hydrochloride 96.0+%, TCI America™

$54.06 - $54.06

Chemical Identifiers

CAS 2491-18-1
Molecular Formula C6H14ClNO2S
Molecular Weight (g/mol) 199.69
MDL Number MFCD00012491,MFCD00067540
InChI Key MEVUPUNLVKELNV-UHFFFAOYNA-N
Synonym h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
PubChem CID 11435579
IUPAC Name hydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride
SMILES [H+].[Cl-].COC(=O)C(N)CCSC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M08535G
View Documents
TCI America
M08535G
5 g
Each for $54.06
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2491-18-1
199.69
MEVUPUNLVKELNV-UHFFFAOYNA-N
11435579
[H+].[Cl-].COC(=O)C(N)CCSC
C6H14ClNO2S
MFCD00012491,MFCD00067540
h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
hydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride

Specifications

2491-18-1
C6H14ClNO2S
5 g
MEVUPUNLVKELNV-UHFFFAOYNA-N
hydrogen methyl 2-amino-4-(methylsulfanyl)butanoate chloride
11435579
≥96.0% (T)
L-Methionine Methyl Ester Hydrochloride
151°C
MFCD00012491,MFCD00067540
h-met-ome.hcl, l-methionine methyl ester hydrochloride, h-met-ome hydrochloride, methionine methyl ester hcl, methyl 2s-2-amino-4-methylsulfanyl butanoate hydrochloride, h-d-met-ome hydrochloride, met-ome.hcl, met-ome. hcl, h-met-ome. hcl, pubchem14925
[H+].[Cl-].COC(=O)C(N)CCSC
199.69
199.69
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'